missing translation for 'onlineSavingsMsg'
Learn More

Troemner™ Individual Analytical Precision Weights, UltraClass with Traceable Certificate

Highly polished kilogram and gram weights

Supplier:  Troemner™ 30390215

 View more versions of this product

Catalog No. 022141K


Only null left
Explore available promotions

Description

Description

Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage.

  • Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F)
  • Fractional weights 500mg to 5mg are made of stainless steel (30mg weights are aluminum); fractional weights 3mg and below are aluminum
  • The UltraClass weight tolerance is equal to ANSI/ASTM E617 Class 0 and exceeds OIML R 111 Class E2
  • This class is used as a reference standard for calibrating other reference standards and weights and is appropriate for calibrating high-precision analytical balances with a readability as low as 0.01mg
  • Weights come in a rugged, precision-fit, high-quality polycarbonate case
  • Supplied with a Traceable Certificate
Specifications

Specifications

Metric Analytical Sheet Weight with One Side Turned Up
UltraClass
500 mg
500 mg
Polycarbonate case
1
Traceable certificate, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Flat with one side turned up
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions