missing translation for 'onlineSavingsMsg'
Learn More

Troemner™ Individual Analytical Precision Weights, UltraClass with Traceable Certificate

Catalog No. 022141A
Change view
Click to view available options
Mass:
1 mg
30 mg
100 mg
300 mg
500 mg
1 g
3 g
5 g
10 g
20 g
30 g
100 g
500 g
1 kg
2 kg
5 kg
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mass
022141A 100 g
022141C 30 g
022141D 20 g
022141E 10 g
022141F 5 g
022141G 3 g
022141J 1 g
022141K 500 mg
022141L 300 mg
022141N 100 mg
022141Q 30 mg
022141X 1 mg
02214076 5 kg
02225536 500 g
02225538 2 kg
02225537 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 022141A Supplier Troemner™ Supplier No. 30390224
Only null left

Highly polished kilogram and gram weights

Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage.

  • Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F)
  • Fractional weights 500mg to 5mg are made of stainless steel (30mg weights are aluminum); fractional weights 3mg and below are aluminum
  • The UltraClass weight tolerance is equal to ANSI/ASTM E617 Class 0 and exceeds OIML R 111 Class E2
  • This class is used as a reference standard for calibrating other reference standards and weights and is appropriate for calibrating high-precision analytical balances with a readability as low as 0.01mg
  • Weights come in a rugged, precision-fit, high-quality polycarbonate case
  • Supplied with a Traceable Certificate

Specifications

Product Type Metric Analytical Weight with Handling Knob
Certifications/Compliance Traceable certificate, ISO9001
Classification UltraClass
Material Stainless Steel
Weight (Metric) 100 g
Density 8.03g/cm3 at 20°C
Mass 100 g
Shape Cylindrical with Handling Knob
Includes Polycarbonate case
Quantity Each
Packaging Quantity 1
Show More Show Less