missing translation for 'onlineSavingsMsg'
Learn More

Troemner™ Individual Analytical Precision Weights, Class 1 with Traceable Certificate

Numéro de catalogue. 023181HT
Change view
Click to view available options
Mass:
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
3 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
5 kg
Density:
2.7g/cm3 at 20°C
7.95g/cm3 at 20°C
Material:
Aluminum
Stainless Steel
Shape:
Cylindrical with Handling Knob
Flat with one side turned up
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Mass Density Material Shape
023181HT 500 g 7.95g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
02215100T 1 kg 7.95g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
022141JT 200 g 7.95g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
023191AT 100 g 7.95g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
023191BT 50 g 7.95g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
023191CT 20 g 7.95g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
023191DT 10 g 7.95g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
023191ET 5 g 7.95g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
023191FT 2 g 7.95g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
023191GT 1 g 7.95g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
023191HT 500 mg 7.95g/cm3 at 20°C Stainless Steel Flat with one side turned up
023191JT 200 mg 7.95g/cm3 at 20°C Stainless Steel Flat with one side turned up
023191KT 100 mg 7.95g/cm3 at 20°C Stainless Steel Flat with one side turned up
023191LT 50 mg 7.95g/cm3 at 20°C Stainless Steel Flat with one side turned up
023191NT 20 mg 7.95g/cm3 at 20°C Stainless Steel Flat with one side turned up
023191PT 10 mg 7.95g/cm3 at 20°C Stainless Steel Flat with one side turned up
023191QT 5 mg 7.95g/cm3 at 20°C Stainless Steel Flat with one side turned up
023191RT 2 mg 2.7g/cm3 at 20°C Aluminum Flat with one side turned up
023191TT 1 mg 2.7g/cm3 at 20°C Aluminum Flat with one side turned up
02214077 3 g 7.95g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
02225539 5 kg 7.95g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Numéro de catalogue. 023181HT Fournisseur Troemner™ Code fournisseur 30391598
Il en reste null

Designed for your most demanding weighing applications

Weights are designed so the specifications on surface finish, markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances decrease in order to enable the weight to maintain its tolerance over time.

  • Precision Analytical Individual Weights are made from the finest materials; manufactured in sizes ranging from 1mg through 50kg
  • Fractional weights 5mg to 500mg are made of stainless steel (30mg weights are aluminum)
  • Fractional weights 3mg and below are made of aluminum
  • Weights 1mg to 2kg are supplied in a high-quality polycarbonate case
  • Used as reference standards as well as calibrating Class I balances as specified in OIML R 76
  • Used to calibrate Class 1 weights
  • Calibrate certain Class I and Class II balances and scales as specified in OIML R 76
  • Can also be used to calibrate Class 2 weights

Spécifications

Product Type Metric Analytical Weight with Handling Knob
Certifications/Compliance Traceable certificate, ISO9001
Classification ASTM Class 1
Material Stainless Steel
Weight (Metric) 500 g
Density 7.95g/cm3 at 20°C
Mass 500 g
Shape Cylindrical with Handling Knob
Includes Polycarbonate case
Quantity Each
Packaging Quantity 1
Afficher plus de résultats Afficher moins de résultats