Learn More
Troemner™ Electric Balance Weight Cases
Designed to securely hold and protect weights sized 2kg through 10g
$95.71 - $177.45
Specifications
| Product Type | Individual Electronic Balance Weight Case |
|---|---|
| Case Material | Polycarbonate |
| For Use With (Application) | Securely holds, protects and transports weights |
| Includes | Handle, a recessed grip, side latches and a color-coded insert |
| Quantity | Each |
| Catalog Number | Mfr. No. | Diameter (English) Outside | For Use With (Equipment) | Height (English) Case | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (English) Outside | For Use With (Equipment) | Height (English) Case | Price | Quantity | |||
|
02-215-526
|
Troemner™
80864228 |
2 in. | 30g weight | 2.5 in. |
Each for $102.41
|
|
|||
|
02-215-528
|
Troemner™
80864230 |
2 in. | 40g weight | 2.5 in. |
Each for $102.41
|
|
|||
|
02-215-529
|
Troemner™
80864231 |
2 in. | 50g weight | 2.5 in. |
Each for $102.41
|
|
|||
|
02-215-525
|
Troemner™
80864227 |
2 in. | 20g weight | 2.5 in. |
Each for $102.41
|
|
|||
|
02-215-530
|
Troemner™
80864232 |
2 in. | 60g weight | 2.5 in. |
Each for $102.41
|
|
|||
|
02-215-532
|
Troemner™
80864234 |
2 in. | 80g weight | 2.5 in. |
Each for $95.71
|
|
|||
|
02-215-531
|
Troemner™
80864233 |
2 in. | 70g weight | 2.5 in. |
Each for $95.71
|
|
|||
|
02-215-533
|
Troemner™
80864235 |
2 in. | 100g weight | 2.5 in. |
Each for $95.71
|
|
|||
|
02-215-536
|
Troemner™
80864238 |
2 in. | 180g weight | 2.5 in. |
Each for $95.71
|
|
|||
|
02-215-537
|
Troemner™
80864239 |
2 in. | 200g weight | 2.5 in. |
Each for $95.71
|
|
|||
|
02-215-534
|
Troemner™
80864236 |
2 in. | 150g weight | 2.5 in. |
Each for $95.71
|
|
|||
|
02-215-538
|
Troemner™
80864240 |
2 in. | 300g weight | 2.5 in. |
Each for $140.82
|
|
|||
|
02-215-535
|
Troemner™
80864237 |
2 in. | 160g weight | 2.5 in. |
Each for $95.71
|
|
|||
|
02-215-541
|
Troemner™
80864243 |
3.5 in. | 600g weight | 4.5 in. |
Each for $140.82
|
|
|||
|
02-215-539
|
Troemner™
80864241 |
2 in. | 400g weight | 2.5 in. |
Each for $140.82
|
|
|||
|
02-215-540
|
Troemner™
80864242 |
3.5 in. | 500g weight | 4.5 in. |
Each for $140.82
|
|
|||
|
02-215-542
|
Troemner™
80864244 |
3.5 in. | 1kg weight | 4.5 in. |
Each for $140.82
|
|
|||
|
02-215-544
|
Troemner™
80864247 |
3.5 in. | 1.5kg weight | 4.5 in. |
Each for $140.82
|
|
|||
|
02-215-543
|
N/A | 3.5 in. | 1.2kg weight | 4.5 in. |
Each for $177.45
|
|
|||
|
02-215-545
|
Troemner™
80864248 |
3.5 in. | 2kg weight | 4.5 in. |
Each for $140.82
|
|
|||
Description
- Lid is rotated open, color-coded insert and weight move with lid
- Prevents the urethane bumper from rubbing on weight, further protecting weight's calibration
Specifications
| Individual Electronic Balance Weight Case | |
| Securely holds, protects and transports weights | |
| Each |
| Polycarbonate | |
| Handle, a recessed grip, side latches and a color-coded insert |