missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIzol™ Reagent and Phasemaker™ Tubes Complete System

This system includes both TRIzol™ Reagent and Invitrogen™ Phasemaker™ Tubes.

$994.00

Specifications

Isolation Technology Organic Extraction
Purification Time 1 hr.
Elution Volume 20 to 50 μL
Sample Type Bacteria, Cells, Plants, Tissue, Yeast
Final Product Type Total RNA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
A33251
Encompass
View Documents Promotion Details
Invitrogen™
A33251
200 Preps
Each for $994.00
Only null left
 
Description

Description

This is a complete system that includes the TRIzol™ Reagent and the Phasemaker™ Tubes. The TRIzol Reagent offers superior lysis capability, even with difficult sample types. Invitrogen™ Phasemaker Tubes contain a polymer that clearly separates and completely isolates the upper aqueous phase of the TRIzol mix from the organic phase underneath, making very easy to pipette off the upper phase containing the RNA.

The polymer works on density gradient, as it is heavier than the aqueous phase, but lighter that the organic phase; after centrifugation it positions itself between these two layers. It creates a tight seal between the two phases, making it very easy to pipette off the aqueous phase without the risk of carrying over contaminants from the organic layer. Because this, Invitrogen Phasemaker Tubes allow you to recover the whole aqueous phase, increasing yield up to 30%. It is an inert polymer that does not interfere with downstream experiments such as PCR. It works with fresh or frozen samples (tissue or blood). It also works with tissue samples treated previously with RNAlater. Invitrogen Phasemaker Tubes come already pre-dispensed. The tube size is 2 mL.

• Complete System includes TRIzol and Phasemaker Tubes
• Phasemaker Tubes are pre-dispensed in 2 mL tubes
• Creates a tight seal between the aqueous and the organic phases
• Easier to pipette off the whole aqueous phase
• Increases yield up to 30%
• RNA integrity is not compromised
• The seal stays for longer periods of time, allowing to increase the number of samples per batch
• Inert, does not interfere with downstream experiments

Warranty

One year after receiving

Specifications

Specifications

Organic Extraction
20 to 50 μL
Total RNA
RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling
Bacteria: ≤107 cells
Cells: ≤107
Plant: ≤107 cells, 100 mg
Tissue: ≤100 mg
Yeast: ≤107 cells
1 hr.
Bacteria, Cells, Plants, Tissue, Yeast
Not High-throughput Compatible (Manual)
Room Temperature
Cells: ≤15 μg
Tissue: ≤73 μg
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.