missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIzol™ LS Reagent and Phasemaker™ Tubes Complete System

Catalog No. A33253
Encompass
Change view
Click to view available options
Quantity:
100 Preps
200 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A33253 200 Preps
A33252 100 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A33253 Supplier Invitrogen™ Supplier No. A33253

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

One bottle of TRIzol LS Reagent 200 mL and two bags of 100 Phasemaker Tubes

This system includes both TRIzol™ LS Reagent and Invitrogen™ Phasemaker™ Tubes.

This is a complete system that includes the TRIzol™ Reagent and the Phasemaker™ Tubes. The TRIzol Reagent offers superior lysis capability, even with difficult sample types. Invitrogen™ Phasemaker Tubes contain a polymer that clearly separates and completely isolates the upper aqueous phase of the TRIzol mix from the organic phase underneath, making very easy to pipette off the upper phase containing the RNA.

The polymer works on density gradient, as it is heavier than the aqueous phase, but lighter that the organic phase; after centrifugation it positions itself between these two layers. It creates a tight seal between the two phases, making it very easy to pipette off the aqueous phase without the risk of carrying over contaminants from the organic layer. Because this, Invitrogen Phasemaker Tubes allow you to recover the whole aqueous phase, increasing yield up to 30%. It is an inert polymer that does not interfere with downstream experiments such as PCR. It works with fresh or frozen samples (tissue or blood). It also works with tissue samples treated previously with RNAlater. Invitrogen Phasemaker Tubes come already pre-dispensed. The tube size is 2 mL.

• Complete System includes TRIzol and Phasemaker Tubes
• Phasemaker Tubes are pre-dispensed in 2 mL tubes
• Creates a tight seal between the aqueous and the organic phases
• Easier to pipette off the whole aqueous phase
• Increases yield up to 30%
• RNA integrity is not compromised
• The seal stays for longer periods of time, allowing to increase the number of samples per batch
• Inert, does not interfere with downstream experiments

Warranty

One year after receiving

Specifications

Content And Storage • 200 mL TRIzol LS Reagent; room temperature
• 200 Phasemaker Tubes; room temperature

Isolation Technology Organic Extraction
Purification Time 1 hr.
Elution Volume 20 to 50 μL
Sample Type Liquid Samples (e.g. Serum)
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling
Quantity 200 Preps
Shipping Condition Room Temperature
Starting Material Amount 0.25 mL per 0.75 mL of TRIzol™ LS Reagent
Yield Tissue: ≤10 μg per mg
Cells: ≤15 μg per 106 cells
Show More Show Less
Do Phasemaker Tubes affect the performance of downstream applications like RT PCR ?

The polymer in Phasemaker Tubes is inert and heat-stable, and does not affect standard downstream applications performed after RNA extraction.

Can I use Phasemaker Tubes with samples that have been pre-treated with RNAlater?

Yes you can. As RNAlater incorporates salts into the sample, the salt content can increase the density of the sample mix, so we recommend adding RNAse-free water (50-100 µL) to the sample mix.

How do Phasemaker Tubes work?

Phasemaker Tubes work on the principle of density gradient centrifugation. Phasemaker Tubes contain a thick liquid polymer that is heavier than the aqueous phase of the TRIzol Reagent mix, but lighter than the organic phase, so after centrifugation, the polymer positions itself between these two layers and forms an exclusive physical barrier. It separates and completely isolates the upper aqueous phase from the organic phase underneath, making it very easy to aspirate the upper phase containing the RNA.

Can Phasemaker Tubes be used with organic reagents other than TRIzol Reagent?

Phasemaker Tubes work exclusively with TRIzol Reagent (and TRIzol LS Reagent ). As other similar reagents may have different density, Phasemaker Tubes may not work properly.

Can Phasemaker Tubes be used for isolation of DNA or protein?

No. Phasemaker tubes are only validated for RNA extraction.

For Research Use Only. Not for use in diagnostic procedures.