missing translation for 'onlineSavingsMsg'
Learn More

Trityl Bromide 98.0+%, TCI America™

Catalog No. t051225g
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T051225G 25 g
T0512100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T051225G Supplier TCI America Supplier No. T051225G
Only null left

Chemical Identifiers

CAS 596-43-0
Molecular Formula C19H15Br
Molecular Weight (g/mol) 323.233
MDL Number MFCD00000120
InChI Key NZHXEWZGTQSYJM-UHFFFAOYSA-N
Synonym bromotriphenylmethane, triphenylmethyl bromide, trityl bromide, triphenylbromomethane, bromodiphenylmethyl benzene, methane, bromotriphenyl, benzene, 1,1',1-bromomethylidyne tris, triphenylmethylbromide, bromomethanetriyl tribenzene, 2-bromo-1,1,1-triphenylethane
PubChem CID 11692
IUPAC Name [bromo(diphenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Br

Specifications

Melting Point 156°C
Quantity 25 g
UN Number 3261
Formula Weight 323.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trityl Bromide
RTECSNumber PA5550000
TSCA Yes