missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tritriacontane 95.0+%, TCI America™
$264.70 - $663.60
Chemical Identifiers
| CAS | 630-05-7 |
|---|---|
| Molecular Formula | C33H68 |
| Molecular Weight (g/mol) | 464.91 |
| MDL Number | MFCD00010187 |
| InChI Key | SUJUOAZFECLBOA-UHFFFAOYSA-N |
| Synonym | n-tritriacontane, n-tritiacontane, tritriacontane, dotriacontane, methyl, acmc-1b412, ch3-ch2 31-ch3, tritriacontane, analytical standard, tritriacontane, purum gc |
| PubChem CID | 12411 |
| ChEBI | CHEBI:9751 |
| IUPAC Name | tritriacontane |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Chemical Identifiers
| 630-05-7 | |
| 464.91 | |
| SUJUOAZFECLBOA-UHFFFAOYSA-N | |
| 12411 | |
| tritriacontane |
| C33H68 | |
| MFCD00010187 | |
| n-tritriacontane, n-tritiacontane, tritriacontane, dotriacontane, methyl, acmc-1b412, ch3-ch2 31-ch3, tritriacontane, analytical standard, tritriacontane, purum gc | |
| CHEBI:9751 | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Specifications
| 630-05-7 | |
| White-Yellow | |
| MFCD00010187 | |
| n-tritriacontane, n-tritiacontane, tritriacontane, dotriacontane, methyl, acmc-1b412, ch3-ch2 31-ch3, tritriacontane, analytical standard, tritriacontane, purum gc | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
| 464.91 | |
| CHEBI:9751 | |
| ≥95.0% (GC) | |
| Tritriacontane |
| 72°C | |
| C33H68 | |
| 1 g | |
| SUJUOAZFECLBOA-UHFFFAOYSA-N | |
| tritriacontane | |
| 12411 | |
| 464.91 | |
| Crystalline Powder |
Safety and Handling
TSCA : No