CAS RN 630-05-7
IUPAC Name:
tritriacontane
Synonyms:
tritriacontane
Molecular Weight (g/mol):
464.91
Molecular Formula:
C33H68
InChi Key:
SUJUOAZFECLBOA-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
1
–
1
of
1
results
1
Tritriacontane 95.0+%, TCI America™
CAS: 630-05-7 Molecular Formula: C33H68 Molecular Weight (g/mol): 464.91 MDL Number: MFCD00010187 InChI Key: SUJUOAZFECLBOA-UHFFFAOYSA-N Synonym: n-tritriacontane,n-tritiacontane,tritriacontane,dotriacontane, methyl,acmc-1b412,ch3-ch2 31-ch3,tritriacontane, analytical standard,tritriacontane, purum gc PubChem CID: 12411 ChEBI: CHEBI:9751 IUPAC Name: tritriacontane SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC