missing translation for 'onlineSavingsMsg'
Learn More

Tritriacontane 95.0+%, TCI America™

Catalog No. T09681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09681G 1 g
T09685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09681G Supplier TCI America Supplier No. T09681G
Only null left

Chemical Identifiers

CAS 630-05-7
Molecular Formula C33H68
Molecular Weight (g/mol) 464.91
MDL Number MFCD00010187
InChI Key SUJUOAZFECLBOA-UHFFFAOYSA-N
Synonym n-tritriacontane, n-tritiacontane, tritriacontane, dotriacontane, methyl, acmc-1b412, ch3-ch2 31-ch3, tritriacontane, analytical standard, tritriacontane, purum gc
PubChem CID 12411
ChEBI CHEBI:9751
IUPAC Name tritriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 1 g
Formula Weight 464.91
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tritriacontane