missing translation for 'onlineSavingsMsg'
Learn More

Triton™ X-114, MP Biomedicals™

Triton™ X-114 is a surfactant used in industrial and household detergent applications and in emulsifying agents.

$82.13

Chemical Identifiers

CAS 9036-19-5
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN19397190
View Documents Promotion Details
N/A 500 mL
Each for $82.13
Only null left
 
Description

Description

  • Triton™ X-114 is used in biochemical applications to solubilize and separate proteins due to its low cloud point (23°C).
  • At temperatures above the cloud point, detergent solutions separate into aqueous and detergent-enriched phases.
  • This makes Triton™ X-114 particularly useful in separating lipophilic proteins from hydrophilic proteins.
Specifications

Chemical Identifiers

9036-19-5

Specifications

Light Yellow
Liquid
Plastic Container
(C2H4O)x · C14H22O (x= 7-8)
Triton™ X-114
9036-19-5
8.8 lb./gal
1.054 g/ml
294.435 g/mol
Videos
Safety and Handling

Safety and Handling

Recommended Storage : 15 to 30°C

SDS
Documents

Documents

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.