missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triton X-100, For Analysis, MilliporeSigma™
$362.21 - $665.22
Chemical Identifiers
| CAS | 9036-19-5 |
|---|---|
| Molecular Formula | C8H17C6H4(OCH2CH2)nOH |
| Synonym | Octoxynol |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
M1086031000
|
N/A | 1 L | Glass Bottle |
Each for $362.21
|
|
||||
|
M1086032500
|
N/A | 2.5 L | Glass Bottle |
Each for $665.22
|
|
||||
Chemical Identifiers
| 9036-19-5 | |
| C8H17C6H4(OCH2CH2)nOH | |
| Octoxynol |
Specifications
| Colorless | |
| Liquid | |
| 1.07 g/3 (20 °C) | |
| 251°C | |
| Octoxynol |
| 9036-19-5 | |
| >200°C (1013hPa) | |
| 5.0 to 8.0 (10 g/L, H2O, 20°C) | |
| C8H17C6H4(OCH2CH2)nOH | |
| Ether |
Safety and Handling
RTECSNumber : MD0907700