missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triton(R) X-405, 70% Solution in Water, Spectrum™ Chemical
$344.41 - $22,268.28
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18614597
|
N/A | 500 mL | Poly Bottle |
Each for $344.41
|
|
||||
|
18614598
|
N/A | 4 L | Poly Bottle |
Each for $1,181.31
|
|
||||
|
18614599
|
N/A | 20 L | Steel Can |
Each for $3,596.31
|
|
||||
|
18614600
|
N/A | 200 L | Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 9036-19-5 |
Specifications
| 9036-19-5 | |
| 1 | |
| 7.0 to 9.0 | |
| Reagent |
CAUTION: For professional use only. Not for household use. Read and understand the label and Safety Data Sheet (SDS) prior to use.