missing translation for 'onlineSavingsMsg'
Learn More

Triton(R) X-405, 70% Solution in Water, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor TR14020LTMT

Quantity 20 L
Packaging Steel Can

 View more versions of this product

Catalog No. 18614599


Only null left

Description

Description

Specifications

Chemical Identifiers

9036-19-5

Specifications

9036-19-5
7.0 to 9.0
Steel Can
1
20 L
Reagent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions