missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triton(R) X-100, Spectrum™ Chemical
TR135, 9036-19-5, C14H21(C2H4O)9-10OH
$1,334.54 - $26,424.21
Chemical Identifiers
| CAS | 9036-19-5 |
|---|---|
| Molecular Formula | C18H30O3 |
| Molecular Weight (g/mol) | 294.44 |
| InChI Key | LBCZOTMMGHGTPH-UHFFFAOYSA-N |
| IUPAC Name | 2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethan-1-ol |
| SMILES | CC(C)(C)CC(C)(C)C1=CC=C(OCCOCCO)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18607597
|
N/A | 4 L | Poly Bottle |
Each for $1,334.54
|
|
||||
|
18607598
|
N/A | 20 L | Steel Can |
Each for $4,200.21
|
|
||||
|
18607599
|
N/A | 200 L | Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Triton X-100
Chemical Identifiers
| 9036-19-5 | |
| 294.44 | |
| 2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethan-1-ol |
| C18H30O3 | |
| LBCZOTMMGHGTPH-UHFFFAOYSA-N | |
| CC(C)(C)CC(C)(C)C1=CC=C(OCCOCCO)C=C1 |
Specifications
| 9036-19-5 | |
| C18H30O3 | |
| CC(C)(C)CC(C)(C)C1=CC=C(OCCOCCO)C=C1 | |
| 294.44 |
| 1 | |
| LBCZOTMMGHGTPH-UHFFFAOYSA-N | |
| 2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethan-1-ol | |
| Reagent |