missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Triton∣r X-100 lysis buffer, pH 7.4

Catalog No. AAJ62289AK
Change view
Click to view available options
Quantity:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62289AK 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62289AK Supplier Thermo Scientific Chemicals Supplier No. J62289AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

It is widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions. Triton X-100 is a commonly used detergent in laboratories. Triton X-100 is widely used to lyse cells to extract protein or organelles, or to permeabilize the membranes of living cells. Apart from laboratory use, Triton X-100 can be found in several types of cleaning compounds,[8] ranging from heavy-duty industrial products to gentle detergents. It is also a popular ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol. It is a good micellar catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Quantity 250mL
Format Liquid
Product Type Cell Lysis Reagent
pH 7.4
Content And Storage 2° to 8°C

RUO – Research Use Only