Learn More
Trithiocyanuric acid, 95%
CAS: 638-16-4 | C3H3N3S3 | 177.258 g/mol
Supplier: Thermo Scientific Chemicals A1908022
Description
Reaction of BunLi with trithiocyanuric acid (LH3) in toluene/HMPA yielded a mono-lithiated species, [(LH2Li)·HMPA2]. Metal complexes of trithiocyanuric acid is used for the elucidation of coordination polymer stoichiometry via thermometric titrimetry:.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReaction of BunLi with trithiocyanuric acid (LH3) in toluene/HMPA yielded a mono-lithiated species, [(LH2Li)·HMPA2]. Metal complexes of trithiocyanuric acid is used for the elucidation of coordination polymer stoichiometry via thermometric titrimetry:.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| Trithiocyanuric acid | |
| >300°C | |
| MFCD00006052 | |
| trithiocyanuric acid, thiocyanuric acid, 1,3,5-triazine-2,4,6-trithiol, trismercaptotriazine, trimercaptocyanuric acid, cyanuric acid, trithio, usaf th-3, 1,3,5-triazine-2,4,6 1h,3h,5h-trithione, s-triazine-2,4,6-trithiol, 2,4,6-triazinetrithiol | |
| WZRRRFSJFQTGGB-UHFFFAOYSA-N | |
| 1,3,5-triazinane-2,4,6-trithione | |
| 1268121 | |
| 95% |
| 638-16-4 | |
| C3H3N3S3 | |
| 100 g | |
| Slightly soluble in water. | |
| C1(=S)NC(=S)NC(=S)N1 | |
| 177.258 | |
| 177.27 |
Chemical Identifiers
| 638-16-4 | |
| 177.258 | |
| WZRRRFSJFQTGGB-UHFFFAOYSA-N | |
| 1268121 | |
| C1(=S)NC(=S)NC(=S)N1 |
| C3H3N3S3 | |
| MFCD00006052 | |
| trithiocyanuric acid, thiocyanuric acid, 1,3,5-triazine-2,4,6-trithiol, trismercaptotriazine, trimercaptocyanuric acid, cyanuric acid, trithio, usaf th-3, 1,3,5-triazine-2,4,6 1h,3h,5h-trithione, s-triazine-2,4,6-trithiol, 2,4,6-triazinetrithiol | |
| 1,3,5-triazinane-2,4,6-trithione |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-322-8
RTECSNumber : XZ2830000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only