missing translation for 'onlineSavingsMsg'
Learn More

Trithiocyanuric acid, 95%

Catalog No. p-7024434
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1908006 5 g
AAA1908014 25 g
AAA1908022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1908006 Supplier Thermo Scientific Chemicals Supplier No. A1908006
Only null left

CAS: 638-16-4 | C3H3N3S3 | 177.258 g/mol

Reaction of BunLi with trithiocyanuric acid (LH3) in toluene/HMPA yielded a mono-lithiated species, [(LH2Li)·HMPA2]. Metal complexes of trithiocyanuric acid is used for the elucidation of coordination polymer stoichiometry via thermometric titrimetry:.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reaction of BunLi with trithiocyanuric acid (LH3) in toluene/HMPA yielded a mono-lithiated species, [(LH2Li)·HMPA2]. Metal complexes of trithiocyanuric acid is used for the elucidation of coordination polymer stoichiometry via thermometric titrimetry:.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 638-16-4
Molecular Formula C3H3N3S3
Molecular Weight (g/mol) 177.258
MDL Number MFCD00006052
InChI Key WZRRRFSJFQTGGB-UHFFFAOYSA-N
Synonym trithiocyanuric acid, thiocyanuric acid, 1,3,5-triazine-2,4,6-trithiol, trismercaptotriazine, trimercaptocyanuric acid, cyanuric acid, trithio, usaf th-3, 1,3,5-triazine-2,4,6 1h,3h,5h-trithione, s-triazine-2,4,6-trithiol, 2,4,6-triazinetrithiol
PubChem CID 1268121
IUPAC Name 1,3,5-triazinane-2,4,6-trithione
SMILES C1(=S)NC(=S)NC(=S)N1

Specifications

Melting Point >300°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 177.27
Percent Purity 95%
Chemical Name or Material Trithiocyanuric acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-322-8
RTECSNumber XZ2830000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only