missing translation for 'onlineSavingsMsg'
Learn More

Tritetradecyl Borate 98.0+%, TCI America™

Catalog No. B209725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B209725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B209725G Supplier TCI America Supplier No. B209725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23162-15-4
Molecular Formula C42H87BO3
Molecular Weight (g/mol) 650.965
MDL Number MFCD06797053
InChI Key WTBVYJWHTHPPKB-UHFFFAOYSA-N
Synonym Boric Acid Tritetradecyl Ester
PubChem CID 64670
IUPAC Name tritetradecyl borate
SMILES B(OCCCCCCCCCCCCCC)(OCCCCCCCCCCCCCC)OCCCCCCCCCCCCCC

Specifications

Color White
Quantity 25 g
Formula Weight 650.97
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tritetradecyl Borate