accessibility menu, dialog, popup

UserName

CH3OBOH3COCH3

CAS RN 23162-15-4

IUPAC Name: tritetradecyl borate
Synonyms: tritetradecyl borate
Molecular Weight (g/mol): 650.97
Molecular Formula: C42H87BO3
InChi Key: WTBVYJWHTHPPKB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCOB(OCCCCCCCCCCCCCC)OCCCCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results