missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa531715
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531715 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531715 Supplier Invitrogen™ Supplier No. PA531715
Only null left

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (93%), Rat (87%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TRIT1 is responsible for the modification of A37 to isopentenyl A37 of both cytosolic and mitochondrial tRNAs.

Specifications

Antigen TRIT1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Trit1
Gene Accession No. Q80UN9, Q9H3H1
Gene Alias 2310075G14Rik; AI314189; AI314635; AV099619; GRO1; hGRO1; IPP transferase; IPPT; IPT; IPTase; isopentenyl-diphosphate:tRNA isopentenyltransferase; MOD5; Trit1; tRNA dimethylallyltransferase; tRNA dimethylallyltransferase, mitochondrial; tRNA isopentenylpyrophosphate transferase; tRNA isopentenyltransferase; tRNA isopentenyltransferase 1; tRNA isopentenyltransferase, mitochondrial
Gene Symbols Trit1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TRIT1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54802, 66966
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less