missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Tristearin | |
| 73°C | |
| C57H110O6 | |
| 250 g | |
| DCXXMTOCNZCJGO-UHFFFAOYSA-N | |
| 2,3-di(octadecanoyloxy)propyl octadecanoate | |
| 11146 | |
| 891.50 | |
| Crystalline Powder |
| 555-43-1 | |
| White-Yellow | |
| MFCD00036230 | |
| tristearin, stearin, glycerol tristearate, glyceryl tristearate, trioctadecanoin, propane-1,2,3-triyl tristearate, hardened oil, stearin, tri, spezialfett 118, stearic triglyceride | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCC | |
| 891.501 | |
| CHEBI:45956 | |
| ≥80.0% (GC) |
Chemical Identifiers
| 555-43-1 | |
| 891.501 | |
| DCXXMTOCNZCJGO-UHFFFAOYSA-N | |
| 11146 | |
| 2,3-di(octadecanoyloxy)propyl octadecanoate |
| C57H110O6 | |
| MFCD00036230 | |
| tristearin, stearin, glycerol tristearate, glyceryl tristearate, trioctadecanoin, propane-1,2,3-triyl tristearate, hardened oil, stearin, tri, spezialfett 118, stearic triglyceride | |
| CHEBI:45956 | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCC |
Safety and Handling
EINECSNumber : (2)-0669
TSCA : Yes