missing translation for 'onlineSavingsMsg'
Learn More

Trisodium Naphthalene-1,3,6-trisulfonate Hydrate 65.0+%, TCI America™

Catalog No. N002125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N002125G 25 g
N0021100G 100 g
N0021500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N002125G Supplier TCI America Supplier No. N002125G

Chemical Identifiers

CAS 5182-30-9
Molecular Formula C10H5Na3O9S3
Molecular Weight (g/mol) 434.29
MDL Number MFCD00003989
InChI Key NJPKYOIXTSGVAN-UHFFFAOYSA-K
Synonym trisodium 1,3,6-naphthalenetrisulfonate, 1,3,6-naphthalenetrisulfonic acid, trisodium salt, naphthalene-1,3,6-trisulfonic acid trisodium salt, trisodium naphthalene-1,3,6-trisulfonate, sodium naphthalene-1,3,6-trisulfonate, trisodium 1,3,6-naphthalene trisulfonate, trisodium naphthalene-1,3,6-trisulphonate, 1,3,6-naphthalenetrisulfonic acid, sodium salt 1:3, 1,3,6-naphthalene trisulfonic acid trisodium salt, trisodium naphthalene trisulfonate
PubChem CID 78855
IUPAC Name trisodium naphthalene-1,3,6-trisulfonate
SMILES [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Quantity 25 g
Formula Weight 434.29
Percent Purity ≥65.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Trisodium Naphthalene-1,3,6-trisulfonate Hydrate
EINECSNumber (4)-0471
TSCA Yes