missing translation for 'onlineSavingsMsg'
Learn More

Trisodium N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetate Dihydrate 98.0+%, TCI America™

Catalog No. H004725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H004725G 25 g
H0047500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H004725G Supplier TCI America Supplier No. H004725G
Only null left

Chemical Identifiers

CAS 139-89-9
Molecular Formula C10H15N2Na3O7
Molecular Weight (g/mol) 344.206
MDL Number MFCD00043341
InChI Key WHNXAQZPEBNFBC-UHFFFAOYSA-K
Synonym trisodium hedta, hamp-ol crystals, perma kleer 80, chemcolox 800, monaquest ica-120, versen-ol, chel dm 41, detarol trisodium salt, caswell no. 487c, hedta-na3
PubChem CID 8772
IUPAC Name trisodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(2-hydroxyethyl)amino]acetate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CCO)CC(=O)[O-].[Na+].[Na+].[Na+]

Specifications

Melting Point 288°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 344.21
Percent Purity ≥98.0% (T)
Chemical Name or Material Trisodium N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetate Dihydrate
EINECSNumber (2)-1268
RTECSNumber MB9188000
TSCA Yes