accessibility menu, dialog, popup

UserName

NaNaNaHONNOOOOOO

CAS RN 139-89-9

IUPAC Name: trisodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(2-hydroxyethyl)amino)acetate
Synonyms: hedta trisodium hedta
Molecular Weight (g/mol): 344.21
Molecular Formula: C10H15N2Na3O7
InChi Key: WHNXAQZPEBNFBC-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].OCCN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results