missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl) Phosphate 98.0+%, TCI America™

Supplier:  TCI America P12485G

 View more versions of this product

Catalog No. P12485G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tris(trimethylsilyl) Phosphate
Yellow
C9H27O4PSi3
5 g
Phosphoric Acid Tris(trimethylsilyl) Ester
C[Si](C)(C)OP(=O)(O[Si](C)(C)C)O[Si](C)(C)C
314.54
314.54
Liquid
10497-05-9
236°C
MFCD00008267
1993
QJMMCGKXBZVAEI-UHFFFAOYSA-N
tritrimethylsilyl phosphate
25317
≥98.0% (GC)

Chemical Identifiers

10497-05-9
314.54
QJMMCGKXBZVAEI-UHFFFAOYSA-N
25317
C[Si](C)(C)OP(=O)(O[Si](C)(C)C)O[Si](C)(C)C
C9H27O4PSi3
MFCD00008267
Phosphoric Acid Tris(trimethylsilyl) Ester
tritrimethylsilyl phosphate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : TC9700000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions