Learn More
Tris(trimethylsilyl) borate, 98%
CAS: 4325-85-3 | C9H27BO3Si3 | 278.377 g/mol
Supplier: Thermo Scientific Chemicals L0627714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tris(trimethylsilyl) borate | |
| -35°C | |
| 183°C to 185°C | |
| C9H27BO3Si3 | |
| MFCD00051588 | |
| UN1993 | |
| Moisture sensitive | |
| YZYKZHPNRDIPFA-UHFFFAOYSA-N | |
| tris(trimethylsilyl) borate | |
| 78020 | |
| 98% |
| 4325-85-3 | |
| 0.834 | |
| 42°C (107°F) | |
| 1.3861 | |
| 25 g | |
| 1772733 | |
| tris trimethylsilyl borate, tris trimethylsiloxy boron, silanol, trimethyl-, triester with boric acid h3bo3, tritrimethylsilyl borate, borane, tris trimethylsiloxy, silanol, 1,1,1-trimethyl-, 1,1',1-triester with boric acid h3bo3, acmc-209jtk, tris trimethylsiloxy borane, boric acid, 3tms derivative | |
| B(O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C | |
| 278.377 | |
| 278.38 |
Chemical Identifiers
| 4325-85-3 | |
| 278.377 | |
| YZYKZHPNRDIPFA-UHFFFAOYSA-N | |
| 78020 | |
| B(O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C |
| C9H27BO3Si3 | |
| MFCD00051588 | |
| tris trimethylsilyl borate, tris trimethylsiloxy boron, silanol, trimethyl-, triester with boric acid h3bo3, tritrimethylsilyl borate, borane, tris trimethylsiloxy, silanol, 1,1,1-trimethyl-, 1,1',1-triester with boric acid h3bo3, acmc-209jtk, tris trimethylsiloxy borane, boric acid, 3tms derivative | |
| tris(trimethylsilyl) borate |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 000-000-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only