missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl) borate, 98%

Catalog No. p-7044409
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
AAL0627706 5 g
AAL0627714 25 g
2 options

Catalog No. AAL0627706

Supplier: Thermo Scientific Chemicals L0627706

Only null left

CAS: 4325-85-3 | C9H27BO3Si3 | 278.377 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4325-85-3
Molecular Formula C9H27BO3Si3
Molecular Weight (g/mol) 278.377
MDL Number MFCD00051588
InChI Key YZYKZHPNRDIPFA-UHFFFAOYSA-N
Synonym tris trimethylsilyl borate, tris trimethylsiloxy boron, silanol, trimethyl-, triester with boric acid h3bo3, tritrimethylsilyl borate, borane, tris trimethylsiloxy, silanol, 1,1,1-trimethyl-, 1,1',1-triester with boric acid h3bo3, acmc-209jtk, tris trimethylsiloxy borane, boric acid, 3tms derivative
PubChem CID 78020
IUPAC Name tris(trimethylsilyl) borate
SMILES B(O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Melting Point -35°C
Density 0.834
Boiling Point 183°C to 185°C
Flash Point 42°C (107°F)
Refractive Index 1.3861
Quantity 5 g
UN Number UN1993
Beilstein 1772733
Sensitivity Moisture sensitive
Formula Weight 278.38
Percent Purity 98%
Chemical Name or Material Tris(trimethylsilyl) borate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only