missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(pentafluorophenyl)phosphine 95.0+%, TCI America™
$100.67 - $303.84
Chemical Identifiers
| CAS | 1259-35-4 |
|---|---|
| Molecular Formula | C18F15P |
| Molecular Weight (g/mol) | 532.15 |
| MDL Number | MFCD00079654 |
| InChI Key | FQLSDFNKTNBQLC-UHFFFAOYSA-N |
| Synonym | tris pentafluorophenyl phosphine, phosphine, tris pentafluorophenyl, tris perfluorophenyl phosphine, tris pentafluorophenyl phosphane, tris 2,3,4,5,6-pentafluorophenyl phosphane, pubchem6422, acmc-1bogg, ksc491k1f, tris-pentafluorophenyl-phosphane |
| PubChem CID | 102076 |
| IUPAC Name | tris(2,3,4,5,6-pentafluorophenyl)phosphane |
| SMILES | FC1=C(F)C(F)=C(P(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F |
Chemical Identifiers
| 1259-35-4 | |
| 532.15 | |
| FQLSDFNKTNBQLC-UHFFFAOYSA-N | |
| 102076 | |
| FC1=C(F)C(F)=C(P(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F |
| C18F15P | |
| MFCD00079654 | |
| tris pentafluorophenyl phosphine, phosphine, tris pentafluorophenyl, tris perfluorophenyl phosphine, tris pentafluorophenyl phosphane, tris 2,3,4,5,6-pentafluorophenyl phosphane, pubchem6422, acmc-1bogg, ksc491k1f, tris-pentafluorophenyl-phosphane | |
| tris(2,3,4,5,6-pentafluorophenyl)phosphane |
Specifications
| 1259-35-4 | |
| White-Yellow | |
| MFCD00079654 | |
| tris pentafluorophenyl phosphine, phosphine, tris pentafluorophenyl, tris perfluorophenyl phosphine, tris pentafluorophenyl phosphane, tris 2,3,4,5,6-pentafluorophenyl phosphane, pubchem6422, acmc-1bogg, ksc491k1f, tris-pentafluorophenyl-phosphane | |
| FC1=C(F)C(F)=C(P(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F | |
| 532.15 | |
| 532.15 | |
| Crystalline Powder |
| 116°C | |
| C18F15P | |
| 1 g | |
| FQLSDFNKTNBQLC-UHFFFAOYSA-N | |
| tris(2,3,4,5,6-pentafluorophenyl)phosphane | |
| 102076 | |
| ≥95.0% (GC) | |
| Tris(pentafluorophenyl)phosphine |
Safety and Handling
TSCA : No