missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(hydroxymethyl)nitromethane, 95%
Description
Tris(hydroxymethyl)nitromethane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tris(hydroxymethyl)nitromethane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a primary and secondary intermediate.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. Stable under recommended storage conditions.
Tris(hydroxymethyl)nitromethane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a primary and secondary intermediate.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. Stable under recommended storage conditions.
Specifications
Specifications
| Melting Point | ∽160°C (decomposition) |
| Linear Formula | (HOCH2)3CNO2 |
| Quantity | 100 g |
| Beilstein | 742427 |
| Merck Index | 14,9753 |
| Solubility Information | It is soluble in water. |
| Formula Weight | 151.12 |
| Percent Purity | 95% |
| Chemical Name or Material | Tris(hydroxymethyl)nitromethane |
Safety and Handling
| Hazard Category | H302+H332 |
| Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
| EINECSNumber | 204-769-5 |
| RTECSNumber | TY7350000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only