Learn More
Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure
CAS: 1185-53-1 | C4H11NO3·HCl | 157.6 g/mol
Supplier: Thermo Scientific Chemicals 228030010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferSpecifications
| Tris(hydroxymethyl)aminomethane hydrochloride | |
| 150.0°C to 153.0°C | |
| 3.5 to 5 (1% soln. at 25°C) | |
| 99+% | |
| C4H11NO3·HCl | |
| MFCD00012590 | |
| tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride | |
| QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride | |
| 93573 | |
| 99+% | |
| Crystalline Powder or Crystals |
| 1185-53-1 | |
| White | |
| Authentic | |
| Plastic bottle | |
| (HOCH2)3CNH2·HCl | |
| 1 kg | |
| (1% in water) Clear colorless | |
| C(C(CO)(CO)N)O.Cl | |
| 157.6 | |
| 157.6 | |
| Extra Pure |
Chemical Identifiers
| 1185-53-1 | |
| 157.6 | |
| QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
| 93573 | |
| C(C(CO)(CO)N)O.Cl |
| C4H11NO3·HCl | |
| MFCD00012590 | |
| tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS Signal Word: Warning
EINECSNumber : 214-684-5
RUO – Research Use Only