missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane hydrochloride, 99+%

Catalog No. AAA1137918
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1137918 50 g
AAA1137930 250 g
AAA113790B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1137918 Supplier Thermo Scientific Chemicals Supplier No. A1137918

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00012590
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Melting Point ∼152°C (decomposition)
pH 3.5 to 5.0
Linear Formula (HOCH2)3CNH2·HCl
Quantity 50 g
Beilstein 3675235
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 157.6
Percent Purity ≥99%
Chemical Name or Material Tris (hydroxymethyl)aminomethane hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-684-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only