missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane hydrochloride, 1M stock soln, pH 8.0

Catalog No. aaj63636k2
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63636K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63636K2 Supplier Thermo Scientific Chemicals Supplier No. J63636K2
Only null left

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

Tris, Hydrochloride is commonly used as a buffer component for phenol extractions of DNA or RNA and a buffering component of separating and stacking gels in the characterization of protein products by SDS-PAGE. Tris, Hydrochloride has also been used with urea as a method for antigen retrieval in immunohistochemistry. In smooth muscles Tris, Hydrochloride has been observed to inhibit motor responses to adrenergic motor nerve stimulation. Also uses include pH control in buffering systems for electrophoresis applications. Tris has been used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00012590
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Color Colorless
Concentration 1M stock soln.
Solubility Information Fully miscible with water.
pH 8.0
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Tris(hydroxymethyl)aminomethane hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only