missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Tris(hydroxymethyl)aminomethane acetate salt, 99%
Chemical Identifiers
| CAS | 6850-28-8 |
|---|---|
| Molecular Formula | C4H11NO3·C2H4O2 |
| Molecular Weight (g/mol) | 181.19 |
| InChI Key | PIEPQKCYPFFYMG-UHFFFAOYSA-N |
| Synonym | tris hydroxymethyl aminomethane acetate, tris acetate, tris acetate salt, 2-amino-2-hydroxymethyl propane-1,3-diol acetate salt, trizma acetate, tris hydroxymethyl aminomethane acetate salt, 2-hydroxy-1,1-bis hydroxymethyl ethyl ammonium acetate, 1,3-dihydroxy-2-hydroxymethyl propan-2-aminium acetate, acetic acid; tris buffer, tris-acetate |
| PubChem CID | 16218782 |
| ChEBI | CHEBI:66869 |
| IUPAC Name | acetic acid;2-amino-2-(hydroxymethyl)propane-1,3-diol |
| SMILES | CC(=O)O.C(C(CO)(CO)N)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC434500250
|
Thermo Scientific™
434500250 |
25g | Plastic bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC434501000
|
Thermo Scientific™
434501000 |
100g | Plastic bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC434505000
|
Thermo Scientific™
434505000 |
500g | Plastic bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Biological bufferChemical Identifiers
| 6850-28-8 | |
| 181.19 | |
| tris hydroxymethyl aminomethane acetate, tris acetate, tris acetate salt, 2-amino-2-hydroxymethyl propane-1,3-diol acetate salt, trizma acetate, tris hydroxymethyl aminomethane acetate salt, 2-hydroxy-1,1-bis hydroxymethyl ethyl ammonium acetate, 1,3-dihydroxy-2-hydroxymethyl propan-2-aminium acetate, acetic acid; tris buffer, tris-acetate | |
| CHEBI:66869 | |
| CC(=O)O.C(C(CO)(CO)N)O |
| C4H11NO3·C2H4O2 | |
| PIEPQKCYPFFYMG-UHFFFAOYSA-N | |
| 16218782 | |
| acetic acid;2-amino-2-(hydroxymethyl)propane-1,3-diol |
Specifications
| 6850-28-8 | |
| 99% | |
| tris hydroxymethyl aminomethane acetate, tris acetate, tris acetate salt, 2-amino-2-hydroxymethyl propane-1,3-diol acetate salt, trizma acetate, tris hydroxymethyl aminomethane acetate salt, 2-hydroxy-1,1-bis hydroxymethyl ethyl ammonium acetate, 1,3-dihydroxy-2-hydroxymethyl propan-2-aminium acetate, acetic acid; tris buffer, tris-acetate | |
| PIEPQKCYPFFYMG-UHFFFAOYSA-N | |
| acetic acid;2-amino-2-(hydroxymethyl)propane-1,3-diol | |
| 16218782 | |
| 181.19 | |
| Tris(hydroxymethyl)aminomethane acetate salt |
| 0.5 max. (c=50, water) at 290nm | |
| C4H11NO3·C2H4O2 | |
| (1g/ml in water) Clear colorless | |
| CC(=O)O.C(C(CO)(CO)N)O | |
| 181.19 | |
| CHEBI:66869 | |
| 99% |
Safety and Handling
EINECSNumber : 229-939-6
RUO – Research Use Only