missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane acetate salt, 99%

Catalog No. AC434500250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434500250 25 g Plastic bottle
AC434501000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434500250 Supplier Thermo Scientific Chemicals Supplier No. 434500250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6850-28-8 | C4H11NO3·C2H4O2 | 181.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 6850-28-8
Molecular Formula C4H11NO3·C2H4O2
Molecular Weight (g/mol) 181.19
InChI Key PIEPQKCYPFFYMG-UHFFFAOYSA-N
Synonym tris hydroxymethyl aminomethane acetate, tris acetate, tris acetate salt, 2-amino-2-hydroxymethyl propane-1,3-diol acetate salt, trizma acetate, tris hydroxymethyl aminomethane acetate salt, 2-hydroxy-1,1-bis hydroxymethyl ethyl ammonium acetate, 1,3-dihydroxy-2-hydroxymethyl propan-2-aminium acetate, acetic acid; tris buffer, tris-acetate
PubChem CID 16218782
ChEBI CHEBI:66869
IUPAC Name acetic acid;2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES CC(=O)O.C(C(CO)(CO)N)O

Specifications

Absorbance 0.5 max. (c=50, water) at 290nm
Color White
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 25 g
Solubility Information (1g/ml in water) Clear colorless
Formula Weight 181.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tris(hydroxymethyl)aminomethane acetate salt
EINECSNumber 229-939-6

RUO – Research Use Only