missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry

Tris(hydroxymethyl)aminomethane, 99+%, C4H11NO3, CAS Number-77-86-1, 2-amino-2-hydroxymethyl propane-1,3-diol, tham, tris, trisamine, tris base, tris buffer, tris hydroxymethyl aminomethane, trizma, trometamol, tromethamine, 2.5kg | CAS: 77-86-1 | C4H11NO3 | 121.14 g/mol

$149.49 - $434.50

Chemical Identifiers

CAS 77-86-1
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC140505000
SDS View Documents
Thermo Scientific Chemicals
140505000
500 g Plastic bottle
Each for $149.49
Only null left
 
AC140500010
SDS View Documents
Thermo Scientific Chemicals
140500010
1 kg Plastic bottle
Each for $246.37
Only null left
 
AC140500025
SDS View Documents
Thermo Scientific Chemicals
140500025
2.5 kg Plastic bottle
Each for $434.50
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

77-86-1
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
MFCD00004679
trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
CHEBI:9754
C(C(CO)(CO)N)O

Specifications

77-86-1
>168.5°C
10.0 to 11.5 (5% aq. soln.)
0.1% max.
99+%
C4H11NO3
MFCD00004679
04, 303
trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
121.14
Biochemistry
Tris(hydroxymethyl)aminomethane
(40% aq. soln.) (1 cm cell), (40% aq. soln.) (1 cm cell) at 0nm, 0.2 max. at 290nm
White
219.0°C to 220.0°C (10.0 mmHg)
Authentic
Plastic bottle
(HOCH2)3CNH2
500 g
15, 9951
Solubility in water: 550g/L (25°C). Other solubilities: 79.1g/L ethylene glycol- 26g/L methanol (25°C),22g/L 95% ethanol- 14g/L dmf (25°C),2g/L acetone- 0.5g/L ethyl acetate (25°C),0.05g/L chloroform- <0.05g/L ccl4 (25°C)
C(C(CO)(CO)N)O
121.14
CHEBI:9754
99+%
Crystalline Powder, Crystals or Chunks
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS Signal Word: Warning

EINECSNumber : 201-064-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only