missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry

Tris(hydroxymethyl)aminomethane, 99+%, C4H11NO3, CAS Number-77-86-1, 2-amino-2-hydroxymethyl propane-1,3-diol, tham, tris, trisamine, tris base, tris buffer, tris hydroxymethyl aminomethane, trizma, trometamol, tromethamine, 2.5kg | CAS: 77-86-1 | C4H11NO3 | 121.14 g/mol

Supplier:  Thermo Scientific Chemicals 140505000

 View more versions of this product

Catalog No. AC140505000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Specifications

Tris(hydroxymethyl)aminomethane
(40% aq. soln.) (1 cm cell), (40% aq. soln.) (1 cm cell) at 0nm, 0.2 max. at 290nm
White
219.0°C to 220.0°C (10.0 mmHg)
Authentic
Plastic bottle
(HOCH2)3CNH2
500 g
15, 9951
Solubility in water: 550g/L (25°C). Other solubilities: 79.1g/L ethylene glycol- 26g/L methanol (25°C),22g/L 95% ethanol- 14g/L dmf (25°C),2g/L acetone- 0.5g/L ethyl acetate (25°C),0.05g/L chloroform- <0.05g/L ccl4 (25°C)
C(C(CO)(CO)N)O
121.14
CHEBI:9754
99+%
Crystalline Powder, Crystals or Chunks
77-86-1
>168.5°C
10.0 to 11.5 (5% aq. soln.)
0.1% max.
99+%
C4H11NO3
MFCD00004679
04, 303
trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
121.14
Biochemistry

Chemical Identifiers

77-86-1
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
MFCD00004679
trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
CHEBI:9754
C(C(CO)(CO)N)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS Signal Word: Warning

EINECSNumber : 201-064-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only