missing translation for 'onlineSavingsMsg'
Learn More

Tris(hexafluoroacetylacetonato)iron(III) 95.0+%, TCI America™

Catalog No. H05551G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H05551G Supplier TCI America Supplier No. H05551G

Chemical Identifiers

CAS 17786-67-3
Molecular Formula C15H3F18FeO6
Molecular Weight (g/mol) 676.999
MDL Number MFCD00059457
InChI Key NBPRJLXRDBDIFS-VRBCMZOBSA-K
Synonym Ferric(III) Hexafluoroacetylacetonate, Hexafluoroacetylacetono Iron(III) Salt, Iron(III) Hexafluoroacetylacetonate
PubChem CID 53384549
IUPAC Name (E)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate;iron(3+)
SMILES C(=C(C(F)(F)F)[O-])C(=O)C(F)(F)F.C(=C(C(F)(F)F)[O-])C(=O)C(F)(F)F.C(=C(C(F)(F)F)[O-])C(=O)C(F)(F)F.[Fe+3]

Specifications

Melting Point 52°C
Quantity 1 g
Formula Weight 677.00
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(hexafluoroacetylacetonato)iron(III)