missing translation for 'onlineSavingsMsg'
Learn More

Tris(ethylenediamine)cobalt chloride trihydrate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 458960250

 View more versions of this product

Catalog No. AC458960250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Tris(ethylenediamine)cobalt chloride trihydrate
Authentic
C6H24Cl3CoN6·3H2O
tris ethylenediamine cobalt iii chloride trihydrate, cobalt 3+ tris ethylenediamine trihydrate trichloride
WBMVTRUQSMZIKW-UHFFFAOYSA-K
cobalt(3+);ethane-1,2-diamine;trichloride;trihydrate
73995044
97%
14883-80-8
96% min. (Argentometry)
25g
Solubility in water: soluble.
C(CN)N.C(CN)N.C(CN)N.O.O.O.[Cl-].[Cl-].[Cl-].[Co+3]
399.64
399.64

Chemical Identifiers

14883-80-8
399.64
tris ethylenediamine cobalt iii chloride trihydrate, cobalt 3+ tris ethylenediamine trihydrate trichloride
cobalt(3+);ethane-1,2-diamine;trichloride;trihydrate
C6H24Cl3CoN6·3H2O
WBMVTRUQSMZIKW-UHFFFAOYSA-K
73995044
C(CN)N.C(CN)N.C(CN)N.O.O.O.[Cl-].[Cl-].[Cl-].[Co+3]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only