accessibility menu, dialog, popup

UserName

NH2H2NNH2H2NNH2H2NH2OH2OH2OClClClCo

CAS RN 14883-80-8

IUPAC Name: cobalt(3+) tris(ethane-1,2-diamine) trihydrate trichloride
Synonyms: cobalt(3+) tris(ethylenediamine) trihydrate trichloride
Molecular Weight (g/mol): 399.63
Molecular Formula: C6H30Cl3CoN6O3
InChi Key: WBMVTRUQSMZIKW-UHFFFAOYSA-K
SMILES: O.O.O.[Cl-].[Cl-].[Cl-].[Co+3].NCCN.NCCN.NCCN
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)