missing translation for 'onlineSavingsMsg'
Learn More

Tris(diethylamino)phosphine 90.0+%, TCI America™

Supplier:  TCI America T130925ML

 View more versions of this product

Catalog No. T130925ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tris(diethylamino)phosphine
Yellow
C12H30N3P
25 mL
tris diethylamino phosphine, hexaethylphosphorous triamide, hexaethyl phosphorous triamide, phosphorous triamide, hexaethyl, hexaethyltriaminophosphine, n,n,n',n',n,n-hexaethylphosphinetriamine, tris n,n-diethylamino phosphine, hexaethylphosphoruo triamide, hexaethylphosphorus triamide, phosphorous triamide, n,n,n',n',n,n-hexaethyl
CCN(CC)P(N(CC)CC)N(CC)CC
247.37
247.37
Liquid
2283-11-6
245°C
MFCD00009041
1993
FDIOSTIIZGWENY-UHFFFAOYSA-N
[bis(diethylamino)phosphanyl]diethylamine
75292
≥90.0% (GC)

Chemical Identifiers

2283-11-6
247.37
FDIOSTIIZGWENY-UHFFFAOYSA-N
75292
CCN(CC)P(N(CC)CC)N(CC)CC
C12H30N3P
MFCD00009041
tris diethylamino phosphine, hexaethylphosphorous triamide, hexaethyl phosphorous triamide, phosphorous triamide, hexaethyl, hexaethyltriaminophosphine, n,n,n',n',n,n-hexaethylphosphinetriamine, tris n,n-diethylamino phosphine, hexaethylphosphoruo triamide, hexaethylphosphorus triamide, phosphorous triamide, n,n,n',n',n,n-hexaethyl
[bis(diethylamino)phosphanyl]diethylamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions