missing translation for 'onlineSavingsMsg'
Learn More

Tris(diethylamino)phosphine 90.0+%, TCI America™

Catalog No. p-7136709
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13095ML 5 mL
T130925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13095ML Supplier TCI America Supplier No. T13095ML
Only null left

Chemical Identifiers

CAS 2283-11-6
Molecular Formula C12H30N3P
Molecular Weight (g/mol) 247.37
MDL Number MFCD00009041
InChI Key FDIOSTIIZGWENY-UHFFFAOYSA-N
Synonym tris diethylamino phosphine, hexaethylphosphorous triamide, hexaethyl phosphorous triamide, phosphorous triamide, hexaethyl, hexaethyltriaminophosphine, n,n,n',n',n,n-hexaethylphosphinetriamine, tris n,n-diethylamino phosphine, hexaethylphosphoruo triamide, hexaethylphosphorus triamide, phosphorous triamide, n,n,n',n',n,n-hexaethyl
PubChem CID 75292
IUPAC Name [bis(diethylamino)phosphanyl]diethylamine
SMILES CCN(CC)P(N(CC)CC)N(CC)CC

Specifications

Color Yellow
Boiling Point 245°C
Quantity 5 mL
UN Number 1993
Formula Weight 247.37
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(diethylamino)phosphine