missing translation for 'onlineSavingsMsg'
Learn More

Tris(dibenzylideneacetone)dipalladium-chloroform adduct, 97%

Catalog No. p-4259083
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC369340010 1 g Glass bottle
AC369340050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC369340010 Supplier Thermo Scientific Chemicals Supplier No. 369340010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52522-40-4 | C52H43Cl3O3Pd2 | 1035.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52522-40-4
Molecular Formula C52H43Cl3O3Pd2
Molecular Weight (g/mol) 1035.10
MDL Number MFCD00075479
InChI Key LNAMMBFJMYMQTO-UHFFFAOYSA-N
Synonym tris dibenzylideneacetone dipalladium-chloroform adduct, tris dibenylideneacetone dipalladium-chloroform, tris dibenzylideneacetone dipalladium 0 chloroform adduct, tris dibenzylideneacetone dipalladium 0-chloroform adduct, tris dibenzylideneacetone dipalladium chloroform adduct, tris dibenzylideneacetone dipalladium-chloroform, pd2 dba 3 chcl3, tris dibenzylideneacetone chloroform-di-palladium 0, tris dibenzylideneacetone dipalladium chloroform complex
PubChem CID 11029508
IUPAC Name chloroform;(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].[Pd].ClC(Cl)Cl.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1

Specifications

Melting Point 131.0°C to 135.0°C
Color Black to Purple
Assay Percent Range 96% min. (ex Pd)
Packaging Glass bottle
Linear Formula (C6H5CH=CHCOCH=CHC6H5)3Pd2·CHCl3
Quantity 1 g
Fieser 14,339; 15,353; 16,372; 17,394
Formula Weight 1035.11
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Tris(dibenzylideneacetone)dipalladium-chloroform adduct
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only