missing translation for 'onlineSavingsMsg'
Learn More

Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform, Pd 20.6%

Catalog No. p-7059599
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4496903 1 g
AA4496906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4496903 Supplier Thermo Scientific Chemicals Supplier No. 04496903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52522-40-4 | C52H43Cl3O3Pd2 | 1035.10 g/mol

Tris(dibenzylideneacetone)dipalladium(0) is used in Synthesis of nanosized palladium phosphides upon interaction with white phosphorous, Preparation of palladium triphenylphosphine carbonyl cluster complexes, Precursor for synthesis of functionalized multiwalled carbon nanotube-palladium complexes used as catalysts for Heck coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52522-40-4
Molecular Formula C52H43Cl3O3Pd2
Molecular Weight (g/mol) 1035.10
MDL Number MFCD00075479
InChI Key LNAMMBFJMYMQTO-UHFFFAOYSA-N
Synonym tris dibenzylideneacetone dipalladium-chloroform adduct, tris dibenylideneacetone dipalladium-chloroform, tris dibenzylideneacetone dipalladium 0 chloroform adduct, tris dibenzylideneacetone dipalladium 0-chloroform adduct, tris dibenzylideneacetone dipalladium chloroform adduct, tris dibenzylideneacetone dipalladium-chloroform, pd2 dba 3 chcl3, tris dibenzylideneacetone chloroform-di-palladium 0, tris dibenzylideneacetone dipalladium chloroform complex
PubChem CID 11029508
IUPAC Name chloroform;(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].[Pd].ClC(Cl)Cl.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1

Specifications

Melting Point 131°C to 135°C
Linear Formula Pd2(C17H14O)3·CHCl3
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information Soluble in organic solvents. Insoluble in water.
Formula Weight 1035.11
Concentration or Composition (by Analyte or Components) Pd 20.6%
Physical Form Powder
Chemical Name or Material Tris (dibenzylideneacetone)dipalladium (0), Complex with chloroform
Hazard Category H302-H315-H320-H351
Hazard Statement GHS H Statement
H351-H373-H315
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Precautionary Statement P201-P202-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only