missing translation for 'onlineSavingsMsg'
Learn More

Tris(dibenzylideneacetone)dipalladium(0), 97%

CAS: 51364-51-3 | C51H42O3Pd2 | 915.73 g/mol

Supplier:  Thermo Scientific Chemicals 318770250

 View more versions of this product

Catalog No. AC318770250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tris(dibenzylideneacetone)dipalladium(0)
152.0°C to 155.0°C
96.0% min.
97%
C51H42O3Pd2
MFCD00013310
tris dibenzylideneacetone dipalladium 0, tris dibenzylideneacetone dipalladium, pd2 dba 3, tris dibezylideneacetone dipalladium, tris dibenzylideneacetone dipalladium o, tris dibenzylideneacetonyl bis-palladium, tris dba, tris 1e,4e-1,5-diphenylpenta-1,4-dien-3-one dipalladium
CYPYTURSJDMMMP-UHFFFAOYSA-N
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
9811564
97%
51364-51-3
Black to Purple
Authentic
Glass bottle
(C6H5CH=CHCOCH=CHC6H5)3Pd2
25 g
Solubility in water: insoluble. Other solubilities: soluble in chloroform, thf, benzene, toluene
[Pd].[Pd].O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1
915.73
915.7
Fine Crystalline Powder

Chemical Identifiers

51364-51-3
915.73
CYPYTURSJDMMMP-UHFFFAOYSA-N
9811564
[Pd].[Pd].O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1
C51H42O3Pd2
MFCD00013310
tris dibenzylideneacetone dipalladium 0, tris dibenzylideneacetone dipalladium, pd2 dba 3, tris dibezylideneacetone dipalladium, tris dibenzylideneacetone dipalladium o, tris dibenzylideneacetonyl bis-palladium, tris dba, tris 1e,4e-1,5-diphenylpenta-1,4-dien-3-one dipalladium
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only