missing translation for 'onlineSavingsMsg'
Learn More

Tris(dibenzylideneacetone)dipalladium(0), 97%

Catalog No. p-4510547
Change view
Click to view available options
Quantity:
500 mg
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC318775000 500 mg Glass bottle
AC318770050 5 g Glass bottle
AC318770250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC318775000 Supplier Thermo Scientific Chemicals Supplier No. 318775000
Only null left

CAS: 51364-51-3 | C51H42O3Pd2 | 915.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51364-51-3
Molecular Formula C51H42O3Pd2
Molecular Weight (g/mol) 915.73
MDL Number MFCD00013310
InChI Key CYPYTURSJDMMMP-UHFFFAOYSA-N
Synonym tris dibenzylideneacetone dipalladium 0, tris dibenzylideneacetone dipalladium, pd2 dba 3, tris dibezylideneacetone dipalladium, tris dibenzylideneacetone dipalladium o, tris dibenzylideneacetonyl bis-palladium, tris dba, tris 1e,4e-1,5-diphenylpenta-1,4-dien-3-one dipalladium
PubChem CID 9811564
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].[Pd].O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1

Specifications

Melting Point 152.0°C to 155.0°C
Color Black to Purple
Elemental Analysis 96.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C6H5CH=CHCOCH=CHC6H5)3Pd2
Quantity 500 mg
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, thf, benzene, toluene
Formula Weight 915.7
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tris(dibenzylideneacetone)dipalladium(0)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only