Learn More
Tris(dibenzylideneacetone)dipalladium(0), 97%
CAS: 51364-51-3 | C51H42O3Pd2 | 915.73 g/mol
Supplier: Thermo Scientific Chemicals 318770050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tris(dibenzylideneacetone)dipalladium(0) | |
| 152.0°C to 155.0°C | |
| 96.0% min. | |
| 97% | |
| C51H42O3Pd2 | |
| MFCD00013310 | |
| tris dibenzylideneacetone dipalladium 0, tris dibenzylideneacetone dipalladium, pd2 dba 3, tris dibezylideneacetone dipalladium, tris dibenzylideneacetone dipalladium o, tris dibenzylideneacetonyl bis-palladium, tris dba, tris 1e,4e-1,5-diphenylpenta-1,4-dien-3-one dipalladium | |
| CYPYTURSJDMMMP-UHFFFAOYSA-N | |
| (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium | |
| 9811564 | |
| 97% |
| 51364-51-3 | |
| Black to Purple | |
| Authentic | |
| Glass bottle | |
| (C6H5CH=CHCOCH=CHC6H5)3Pd2 | |
| 5 g | |
| Solubility in water: insoluble. Other solubilities: soluble in chloroform, thf, benzene, toluene | |
| [Pd].[Pd].O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1 | |
| 915.73 | |
| 915.7 | |
| Fine Crystalline Powder |
Chemical Identifiers
| 51364-51-3 | |
| 915.73 | |
| CYPYTURSJDMMMP-UHFFFAOYSA-N | |
| 9811564 | |
| [Pd].[Pd].O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1 |
| C51H42O3Pd2 | |
| MFCD00013310 | |
| tris dibenzylideneacetone dipalladium 0, tris dibenzylideneacetone dipalladium, pd2 dba 3, tris dibezylideneacetone dipalladium, tris dibenzylideneacetone dipalladium o, tris dibenzylideneacetonyl bis-palladium, tris dba, tris 1e,4e-1,5-diphenylpenta-1,4-dien-3-one dipalladium | |
| (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
GHS Signal Word: Warning
RUO – Research Use Only