missing translation for 'onlineSavingsMsg'
Learn More

Tris(acetylacetonato)(1,10-phenanthroline)terbium(III) 98.0+%, TCI America™

Catalog No. t3207200mg
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3207200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3207200MG Supplier TCI America Supplier No. T3207200MG

Chemical Identifiers

CAS 18078-86-9
Molecular Formula C27H29N2O6Tb
Molecular Weight (g/mol) 636.462
InChI Key YUDOUPKNKVFAMM-XUHIWKAKSA-K
PubChem CID 91972107
IUPAC Name (Z)-4-oxopent-2-en-2-olate;1,10-phenanthroline;terbium(3+)
SMILES CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[Tb+3]

Specifications

Color White
Quantity 200 mg
Formula Weight 636.46
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(acetylacetonato)(1,10-phenanthroline)terbium(III)