missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris, 99 to 99.5%, MP Biomedicals™
$228.29 - $1,978.04
Chemical Identifiers
| CAS | 77-86-1 |
|---|---|
| Molecular Formula | C4H11NO3 |
| Molecular Weight (g/mol) | 121.136 |
| InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
| Synonym | trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base |
| PubChem CID | 6503 |
| ChEBI | CHEBI:9754 |
| IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
| SMILES | C(C(CO)(CO)N)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15217601
|
MP Biomedicals Inc
0215217601 |
1 kg |
N/A
|
|
|||||
|
ICN15217605
|
MP Biomedicals Inc
0215217605 |
5 kg |
N/A
|
|
|||||
|
ICN15217610
|
MP Biomedicals Inc
0215217610 |
10 kg |
N/A
|
|
|||||
Chemical Identifiers
| 77-86-1 | |
| 121.136 | |
| trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base | |
| CHEBI:9754 | |
| C(C(CO)(CO)N)O |
| C4H11NO3 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 6503 | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol |
Specifications
| 77-86-1 | |
| 99 to 99.5% | |
| 1 kg | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol | |
| 6503 | |
| 121.14 | |
| Tris |
| White | |
| C4H11NO3 | |
| trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base | |
| C(C(CO)(CO)N)O | |
| 121.136 | |
| CHEBI:9754 | |
| Crystalline Powder |