missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris, 99 to 99.5%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0215217605
Specifications
| Tris | |
| White | |
| C4H11NO3 | |
| trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base | |
| C(C(CO)(CO)N)O | |
| 121.136 | |
| CHEBI:9754 | |
| Crystalline Powder |
| 77-86-1 | |
| 99 to 99.5% | |
| 5 kg | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol | |
| 6503 | |
| 121.14 |
Chemical Identifiers
| 77-86-1 | |
| 121.136 | |
| trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base | |
| CHEBI:9754 | |
| C(C(CO)(CO)N)O |
| C4H11NO3 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 6503 | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol |